The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013318.3 | NP_037450.2 | 9 - 10 | 1269 | 70 |
GenBank mRNA | AK309879.1 | - | 2 - 3 | 218 | 70 |
CR749818.1 | - | 3 - 4 | 344 | 70 |