The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013318.3 | NP_037450.2 | 11 - 12 | 1774 | 68 |
GenBank mRNA | AK309879.1 | - | 4 - 5 | 720 | 68 |
CR749818.1 | - | 5 - 6 | 849 | 68 |