The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013318.3 | NP_037450.2 | 15 - 16 | 4454 | 91 |
GenBank mRNA | BX647528.1 | - | 2 - 3 | 2093 | 91 |
BX647842.1 | - | 1 - 2 | 157 | 91 | |
CR749818.1 | - | 10 - 11 | 3529 | 91 | |
CR933661.1 | - | 3 - 4 | 2341 | 91 |