Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZMAT2
Entrez Gene ID
153527
Gene Name
zinc finger matrin-type 2
Gene Aliases
Ptg-12, Snu23
Chromosome Location
Chr.5: 140700360 - 140706681 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144723.2 NP_653324.1 4 - 5 416 88
GenBank mRNA AK055683.1 - 4 - 5 329 88
BC056668.1 - 4 - 5 333 88
BP210985.1 - 4 - 5 410 88
BP370587.1 - 3 - 4 272 88
X97303.1 - 1 - 2 56 88

View Full Product Details