The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177310.1 | NP_001170781.1 | 28 - 29 | 4964 | 159 |
NM_001177311.1 | NP_001170782.1 | 28 - 29 | 4991 | 159 | |
NM_017662.4 | NP_060132.3 | 28 - 29 | 5145 | 159 | |
XM_011518244.1 | XP_011516546.1 | 28 - 29 | 5145 | 159 | |
XM_011518245.1 | XP_011516547.1 | 27 - 28 | 5052 | 159 | |
XM_011518246.1 | XP_011516548.1 | 28 - 29 | 5145 | 159 | |
XM_011518247.1 | XP_011516549.1 | 27 - 28 | 5016 | 159 | |
XM_011518248.1 | XP_011516550.1 | 27 - 28 | 5004 | 159 | |
XM_011518249.1 | XP_011516551.1 | 26 - 27 | 4911 | 159 | |
XM_011518250.1 | XP_011516552.1 | 26 - 27 | 4869 | 159 | |
XM_011518251.2 | XP_011516553.1 | 23 - 24 | 4396 | 159 | |
XM_011518252.2 | XP_011516554.1 | 28 - 29 | 5145 | 159 | |
XM_017014287.1 | XP_016869776.1 | 25 - 26 | 4782 | 159 | |
XM_017014288.1 | XP_016869777.1 | 24 - 25 | 4635 | 159 | |
GenBank mRNA | AF350881.2 | - | 28 - 29 | 5145 | 159 |
AF448232.1 | - | 28 - 29 | 4907 | 159 | |
AK000094.1 | - | 5 - 6 | 1871 | 159 | |
AK225207.1 | - | 5 - 6 | 1871 | 159 | |
AY333282.1 | - | 28 - 29 | 4970 | 159 | |
AY333283.1 | - | 28 - 29 | 4964 | 159 | |
AY333284.1 | - | 28 - 29 | 4991 | 159 | |
AY333285.1 | - | 28 - 29 | 4970 | 159 | |
AY333286.1 | - | 15 - 16 | 1823 | 159 |