Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPM6
Entrez Gene ID
140803
Gene Name
transient receptor potential cation channel subfamily M member 6
Gene Aliases
CHAK2, HMGX, HOMG, HOMG1, HSH
Chromosome Location
Chr.9: 74722495 - 74888094 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177310.1 NP_001170781.1 38 - 39 5987 111
NM_001177311.1 NP_001170782.1 38 - 39 6014 111
NM_017662.4 NP_060132.3 38 - 39 6168 111
XM_011518244.1 XP_011516546.1 38 - 39 6165 111
XM_011518245.1 XP_011516547.1 37 - 38 6075 111
XM_011518246.1 XP_011516548.1 39 - 40 6045 111
XM_011518247.1 XP_011516549.1 37 - 38 6039 111
XM_011518248.1 XP_011516550.1 37 - 38 6027 111
XM_011518249.1 XP_011516551.1 36 - 37 5934 111
XM_011518250.1 XP_011516552.1 36 - 37 5892 111
XM_011518251.2 XP_011516553.1 33 - 34 5419 111
XM_017014287.1 XP_016869776.1 35 - 36 5805 111
XM_017014288.1 XP_016869777.1 34 - 35 5658 111
GenBank mRNA AF350881.2 - 38 - 39 6168 111
AF448232.1 - 38 - 39 5930 111
AK000094.1 - 15 - 16 2894 111
AK225207.1 - 15 - 16 2894 111
AY333282.1 - 38 - 39 5993 111
AY333283.1 - 38 - 39 5987 111
AY333284.1 - 38 - 39 6014 111
AY333285.1 - 39 - 40 6089 111
AY333286.1 - 25 - 26 2846 111
AY333287.1 - 22 - 23 2492 111
AY333288.1 - 12 - 13 1634 111

View Full Product Details