The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003268.5 | NP_003259.2 | 5 - 6 | 636 | 96 |
XM_005273241.4 | XP_005273298.2 | 5 - 6 | 582 | 96 | |
XM_005273242.4 | XP_005273299.2 | 6 - 7 | 702 | 96 | |
XM_005273243.4 | XP_005273300.2 | 4 - 5 | 488 | 96 | |
XM_006711504.3 | XP_006711567.1 | 4 - 5 | 527 | 96 | |
XM_006711505.3 | XP_006711568.1 | 3 - 4 | 433 | 96 | |
XM_006711506.3 | XP_006711569.1 | 2 - 3 | 235 | 96 | |
XM_011509937.2 | XP_011508239.1 | 5 - 6 | 643 | 96 | |
XM_017002208.1 | XP_016857697.1 | 4 - 5 | 549 | 96 | |
GenBank mRNA | AB060695.1 | - | 5 - 6 | 698 | 96 |
AF051151.1 | - | 5 - 6 | 698 | 96 | |
BC109118.1 | - | 3 - 4 | 333 | 96 | |
BC109119.1 | - | 3 - 4 | 333 | 96 |