Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VIT
Entrez Gene ID
5212
Gene Name
vitrin
Gene Aliases
-
Chromosome Location
Chr.2: 36696685 - 36814794 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177969.1 NP_001171440.1 2 - 3 351 100
NM_001177970.1 NP_001171441.1 2 - 3 351 100
NM_001177971.1 NP_001171442.1 2 - 3 351 100
NM_001177972.1 NP_001171443.1 2 - 3 351 100
NM_053276.3 NP_444506.2 2 - 3 351 100
XM_011532905.1 XP_011531207.1 2 - 3 248 100
XM_011532906.1 XP_011531208.1 2 - 3 248 100
XM_017004325.1 XP_016859814.1 2 - 3 356 100
XM_017004326.1 XP_016859815.1 2 - 3 248 100
GenBank mRNA AF063833.1 - 2 - 3 175 100
AK056772.1 - 2 - 3 270 100
AK292149.1 - 2 - 3 248 100
AK299426.1 - 2 - 3 334 100
AL832679.1 - 2 - 3 194 100
AY358338.1 - 2 - 3 236 100
BC061519.1 - 2 - 3 277 100
BC128260.1 - 2 - 3 164 100

View Full Product Details