Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRBN
Entrez Gene ID
51185
Gene Name
cereblon
Gene Aliases
MRT2, MRT2A
Chromosome Location
Chr.3: 3148490 - 3179717 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011533791.2 XP_011532093.1 9 - 10 1030 92

View Full Product Details