Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHLDB3
Entrez Gene ID
653583
Gene Name
pleckstrin homology like domain family B member 3
Gene Aliases
-
Chromosome Location
Chr.19: 43474341 - 43504916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198850.3 NP_942147.3 11 - 12 1602 66
XM_005259169.3 XP_005259226.1 11 - 12 1670 66
XM_005259170.3 XP_005259227.1 11 - 12 1670 66
XM_005259171.3 XP_005259228.1 11 - 12 1670 66
XM_011527236.2 XP_011525538.1 11 - 12 1774 66
GenBank mRNA AA621318.1 - 3 - 4 66
BC157852.1 - 5 - 6 586 66
BC171853.1 - 5 - 6 595 66
BG403419.1 - 5 - 6 453 66

View Full Product Details