Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHLDB3
Entrez Gene ID
653583
Gene Name
pleckstrin homology like domain family B member 3
Gene Aliases
-
Chromosome Location
Chr.19: 43474341 - 43504916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198850.3 NP_942147.3 1 - 2 247 127
XM_005259169.3 XP_005259226.1 1 - 2 330 127
XM_005259170.3 XP_005259227.1 1 - 2 330 127
XM_005259171.3 XP_005259228.1 1 - 2 330 127
GenBank mRNA AK097512.1 - 1 - 2 57 127
BC029299.1 - 1 - 2 247 127
BC070094.1 - 1 - 2 349 127

View Full Product Details