The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198850.3 | NP_942147.3 | 4 - 5 | 794 | 77 |
XM_005259169.3 | XP_005259226.1 | 4 - 5 | 877 | 77 | |
XM_005259170.3 | XP_005259227.1 | 4 - 5 | 877 | 77 | |
XM_005259171.3 | XP_005259228.1 | 4 - 5 | 877 | 77 | |
XM_011527236.2 | XP_011525538.1 | 4 - 5 | 981 | 77 | |
GenBank mRNA | BC070094.1 | - | 4 - 5 | 896 | 77 |
BU858992.1 | - | 2 - 3 | 318 | 77 |