Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHLDB3
Entrez Gene ID
653583
Gene Name
pleckstrin homology like domain family B member 3
Gene Aliases
-
Chromosome Location
Chr.19: 43474341 - 43504916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198850.3 NP_942147.3 6 - 7 1081 83
GenBank mRNA BU858992.1 - 4 - 5 605 83

View Full Product Details