Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LY9
Entrez Gene ID
4063
Gene Name
lymphocyte antigen 9
Gene Aliases
CD229, SLAMF3, hly9, mLY9
Chromosome Location
Chr.1: 160796074 - 160828256 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261456.1 NP_001248385.1 7 - 8 1607 100
XM_011509560.1 XP_011507862.1 7 - 8 1397 100
XM_017001298.1 XP_016856787.1 8 - 9 1667 100
XM_017001299.1 XP_016856788.1 8 - 9 1667 100
XM_017001303.1 XP_016856792.1 7 - 8 1397 100
GenBank mRNA AF055709.1 - 2 - 3 68 100
BC062589.1 - 4 - 5 660 100
L42621.1 - 7 - 8 1400 100

View Full Product Details