Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYMPK
Entrez Gene ID
8189
Gene Name
symplekin
Gene Aliases
SPK, SYM
Chromosome Location
Chr.19: 45815442 - 45863290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004819.2 NP_004810.2 18 - 19 2737 173
XM_005259286.1 XP_005259343.1 18 - 19 2690 173
XM_011527354.1 XP_011525656.1 19 - 20 2740 173
GenBank mRNA AB209024.1 - 18 - 19 4900 173
AB210010.1 - 18 - 19 2582 173
AK130602.1 - 9 - 10 876 173
AK300252.1 - 18 - 19 2591 173
BC006536.2 - 17 - 18 2444 173
BC006567.2 - 17 - 18 2444 173
BX649165.1 - 17 - 18 3643 173
U49240.1 - 13 - 14 2206 173
Y10931.1 - 18 - 19 2555 173

View Full Product Details