Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SERPINF2
Entrez Gene ID
5345
Gene Name
serpin family F member 2
Gene Aliases
A2AP, AAP, ALPHA-2-PI, API, PLI
Chromosome Location
Chr.17: 1742808 - 1755265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000934.3 NP_000925.2 3 - 4 185 66
NM_001165920.1 NP_001159392.1 3 - 4 163 66
NM_001165921.1 NP_001159393.1 3 - 4 185 66
XM_005256701.4 XP_005256758.2 3 - 4 257 66
XM_017024765.1 XP_016880254.1 4 - 5 272 66
GenBank mRNA AK303763.1 - 3 - 4 160 66
BC031592.1 - 3 - 4 163 66
BX107460.1 - 3 - 4 185 66
D00174.1 - 2 - 3 66
J02654.1 - 2 - 3 99 66

View Full Product Details