The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303422.1 | NP_001290351.1 | 12 - 13 | 1384 | 109 |
NM_004490.2 | NP_004481.2 | 13 - 14 | 2013 | 109 | |
XM_017003899.1 | XP_016859388.1 | 13 - 14 | 1450 | 109 | |
GenBank mRNA | AK301961.1 | - | 12 - 13 | 1329 | 109 |
AK313153.1 | - | 13 - 14 | 1642 | 109 | |
BC053559.1 | - | 13 - 14 | 1483 | 109 | |
L76687.1 | - | 13 - 14 | 2012 | 109 |