Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBFOX2
Entrez Gene ID
23543
Gene Name
RNA binding protein, fox-1 homolog 2
Gene Aliases
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
Chromosome Location
Chr.22: 35738736 - 36028892 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031695.2 NP_001026865.1 2 - 3 612 65
NM_001082576.1 NP_001076045.1 2 - 3 612 65
NM_001082577.1 NP_001076046.1 2 - 3 612 65
NM_001082579.1 NP_001076048.1 3 - 4 569 65
XM_005261429.2 XP_005261486.1 3 - 4 924 65
XM_005261432.2 XP_005261489.1 3 - 4 924 65
XM_005261435.1 XP_005261492.1 2 - 3 616 65
XM_006724186.2 XP_006724249.1 3 - 4 924 65
XM_006724190.1 XP_006724253.1 3 - 4 314 65
XM_006724194.2 XP_006724257.1 2 - 3 391 65
XM_017028688.1 XP_016884177.1 3 - 4 924 65
XM_017028689.1 XP_016884178.1 3 - 4 392 65
XM_017028691.1 XP_016884180.1 2 - 3 366 65
XM_017028692.1 XP_016884181.1 3 - 4 661 65
XM_017028694.1 XP_016884183.1 2 - 3 616 65
XM_017028695.1 XP_016884184.1 3 - 4 376 65
XM_017028697.1 XP_016884186.1 3 - 4 1396 65
XM_017028698.1 XP_016884187.1 2 - 3 616 65
GenBank mRNA AB649123.1 - 2 - 3 247 65
AF229058.1 - 2 - 3 417 65
AK055213.1 - 3 - 4 533 65
AK314632.1 - 2 - 3 431 65
AM419009.1 - 2 - 3 271 65
AY072786.1 - 3 - 4 382 65
AY960684.1 - 2 - 3 247 65
BC013115.1 - 2 - 3 436 65
BC025281.1 - 2 - 3 435 65
CR456559.1 - 2 - 3 313 65

View Full Product Details