Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBFOX2
Entrez Gene ID
23543
Gene Name
RNA binding protein, fox-1 homolog 2
Gene Aliases
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
Chromosome Location
Chr.22: 35738736 - 36028892 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082578.1 NP_001076047.1 2 - 3 354 82
NM_001082579.1 NP_001076048.1 2 - 3 354 82
XM_005261428.2 XP_005261485.1 2 - 3 709 82
XM_005261429.2 XP_005261486.1 2 - 3 709 82
XM_005261430.2 XP_005261487.1 2 - 3 709 82
XM_005261431.2 XP_005261488.1 2 - 3 709 82
XM_005261432.2 XP_005261489.1 2 - 3 709 82
XM_005261433.2 XP_005261490.1 2 - 3 709 82
XM_006724185.2 XP_006724248.1 2 - 3 709 82
XM_006724186.2 XP_006724249.1 2 - 3 709 82
XM_006724187.2 XP_006724250.1 2 - 3 709 82
XM_006724188.2 XP_006724251.1 2 - 3 709 82
XM_006724189.3 XP_006724252.1 2 - 3 402 82
XM_006724190.1 XP_006724253.1 2 - 3 99 82
XM_006724193.3 XP_006724256.1 2 - 3 709 82
XM_017028686.1 XP_016884175.1 2 - 3 88 82
XM_017028687.1 XP_016884176.1 2 - 3 269 82
XM_017028688.1 XP_016884177.1 2 - 3 709 82
XM_017028689.1 XP_016884178.1 2 - 3 177 82
XM_017028692.1 XP_016884181.1 2 - 3 446 82
XM_017028695.1 XP_016884184.1 2 - 3 161 82
XM_017028697.1 XP_016884186.1 2 - 3 1181 82
GenBank mRNA AM419009.1 - 1 - 2 56 82
AY072786.1 - 2 - 3 167 82
DB461443.1 - 3 - 4 391 82
DQ778625.1 - 2 - 3 242 82

View Full Product Details