Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNAZ
Entrez Gene ID
2781
Gene Name
G protein subunit alpha z
Gene Aliases
-
Chromosome Location
Chr.22: 23070483 - 23125037 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002073.2 NP_002064.1 1 - 2 94 64
GenBank mRNA BC026342.1 - 1 - 2 94 64
BC037333.1 - 1 - 2 232 64

View Full Product Details