Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMTN
Entrez Gene ID
6525
Gene Name
smoothelin
Gene Aliases
-
Chromosome Location
Chr.22: 31081296 - 31104624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207017.1 NP_001193946.1 16 - 17 2409 71
NM_001207018.1 NP_001193947.1 14 - 15 2338 71
NM_006932.4 NP_008863.3 15 - 16 2334 71
NM_134269.2 NP_599031.1 15 - 16 2334 71
NM_134270.2 NP_599032.2 15 - 16 2334 71
XM_006724298.1 XP_006724361.1 16 - 17 2405 71
XM_006724302.1 XP_006724365.1 16 - 17 2464 71
XM_011530333.2 XP_011528635.1 17 - 18 2510 71
XM_011530341.1 XP_011528643.1 9 - 10 1342 71
XM_017028913.1 XP_016884402.1 17 - 18 2511 71
XM_017028914.1 XP_016884403.1 16 - 17 2417 71
XM_017028915.1 XP_016884404.1 16 - 17 2417 71
XM_017028916.1 XP_016884405.1 17 - 18 2511 71
XM_017028917.1 XP_016884406.1 17 - 18 2511 71
GenBank mRNA AF064238.3 - 15 - 16 2311 71
AJ010306.2 - 15 - 16 2311 71
AK128627.1 - 15 - 16 3972 71
AK295418.1 - 14 - 15 2338 71
AK301289.1 - 5 - 6 610 71
AK304084.1 - 16 - 17 2409 71
AY061971.1 - 15 - 16 2312 71
AY061972.1 - 15 - 16 2312 71
BC034237.2 - 15 - 16 2302 71
Y13492.2 - 15 - 16 2312 71
Z49989.2 - 6 - 7 768 71

View Full Product Details