Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMTN
Entrez Gene ID
6525
Gene Name
smoothelin
Gene Aliases
-
Chromosome Location
Chr.22: 31081296 - 31104624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207017.1 NP_001193946.1 4 - 5 515 74
NM_001207018.1 NP_001193947.1 3 - 4 537 74
NM_006932.4 NP_008863.3 4 - 5 533 74
NM_134269.2 NP_599031.1 4 - 5 533 74
NM_134270.2 NP_599032.2 4 - 5 533 74
XM_006724298.1 XP_006724361.1 4 - 5 511 74
XM_006724302.1 XP_006724365.1 4 - 5 570 74
XM_011530333.2 XP_011528635.1 5 - 6 616 74
XM_011530336.2 XP_011528638.1 5 - 6 617 74
XM_011530337.2 XP_011528639.1 5 - 6 617 74
XM_011530338.2 XP_011528640.1 5 - 6 616 74
XM_011530339.2 XP_011528641.1 5 - 6 617 74
XM_011530340.2 XP_011528642.1 5 - 6 617 74
XM_017028913.1 XP_016884402.1 5 - 6 617 74
XM_017028914.1 XP_016884403.1 5 - 6 616 74
XM_017028915.1 XP_016884404.1 5 - 6 616 74
XM_017028916.1 XP_016884405.1 5 - 6 617 74
XM_017028917.1 XP_016884406.1 5 - 6 617 74
XM_017028918.1 XP_016884407.1 5 - 6 617 74
XM_017028919.1 XP_016884408.1 5 - 6 616 74
GenBank mRNA AB209643.1 - 1 - 2 106 74
AF064238.3 - 4 - 5 510 74
AJ010306.2 - 4 - 5 510 74
AK096953.1 - 4 - 5 568 74
AK128627.1 - 3 - 4 2078 74
AK128719.1 - 4 - 5 379 74
AK295418.1 - 3 - 4 537 74
AK303424.1 - 3 - 4 312 74
AK304084.1 - 4 - 5 515 74
AY061971.1 - 4 - 5 511 74
AY061972.1 - 4 - 5 511 74
BC034237.2 - 4 - 5 501 74
Y13492.2 - 4 - 5 511 74

View Full Product Details