Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH7
Entrez Gene ID
56171
Gene Name
dynein axonemal heavy chain 7
Gene Aliases
-
Chromosome Location
Chr.2: 195737604 - 196075188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018897.2 NP_061720.2 36 - 37 6056 145
XM_011511487.2 XP_011509789.1 37 - 38 6432 145
XM_011511488.2 XP_011509790.1 35 - 36 7106 145
XM_011511489.2 XP_011509791.1 35 - 36 6018 145
XM_011511490.2 XP_011509792.1 35 - 36 7001 145
XM_011511491.2 XP_011509793.1 36 - 37 7228 145
XM_011511492.2 XP_011509794.1 36 - 37 7228 145
XM_011511493.2 XP_011509795.1 36 - 37 7228 145
XM_011511494.2 XP_011509796.1 36 - 37 7227 145
XM_011511495.2 XP_011509797.1 36 - 37 7227 145
XM_011511497.2 XP_011509799.1 3 - 4 452 145
XM_017004504.1 XP_016859993.1 35 - 36 6952 145
GenBank mRNA AB023161.2 - 36 - 37 6074 145
AF327442.1 - 36 - 37 6051 145

View Full Product Details