Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH7
Entrez Gene ID
56171
Gene Name
dynein axonemal heavy chain 7
Gene Aliases
-
Chromosome Location
Chr.2: 195737604 - 196075188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018897.2 NP_061720.2 45 - 46 8700 73
XM_011511487.2 XP_011509789.1 46 - 47 9076 73
XM_011511488.2 XP_011509790.1 44 - 45 9750 73
XM_011511489.2 XP_011509791.1 44 - 45 8662 73
XM_011511490.2 XP_011509792.1 44 - 45 9645 73
XM_011511491.2 XP_011509793.1 45 - 46 9872 73
XM_011511492.2 XP_011509794.1 45 - 46 9872 73
XM_011511493.2 XP_011509795.1 45 - 46 9872 73
XM_011511494.2 XP_011509796.1 45 - 46 9871 73
XM_011511497.2 XP_011509799.1 12 - 13 3096 73
XM_017004504.1 XP_016859993.1 44 - 45 9596 73
GenBank mRNA AB023161.2 - 45 - 46 8718 73
AF327442.1 - 45 - 46 8695 73

View Full Product Details