Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH7
Entrez Gene ID
56171
Gene Name
dynein axonemal heavy chain 7
Gene Aliases
-
Chromosome Location
Chr.2: 195737604 - 196075188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018897.2 NP_061720.2 63 - 64 11868 130
XM_011511487.2 XP_011509789.1 64 - 65 12244 130
XM_011511488.2 XP_011509790.1 62 - 63 12918 130
XM_011511489.2 XP_011509791.1 62 - 63 11830 130
XM_011511490.2 XP_011509792.1 62 - 63 12813 130
XM_011511497.2 XP_011509799.1 30 - 31 6264 130
XM_017004504.1 XP_016859993.1 62 - 63 12764 130
GenBank mRNA AB023161.2 - 63 - 64 11886 130
AF327442.1 - 63 - 64 11863 130
BC029567.1 - 8 - 9 1282 130

View Full Product Details