Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RELN
Entrez Gene ID
5649
Gene Name
reelin
Gene Aliases
ETL7, LIS2, PRO1598, RL
Chromosome Location
Chr.7: 103471784 - 103989516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005045.3 NP_005036.2 3 - 4 631 80
NM_173054.2 NP_774959.1 3 - 4 631 80
GenBank mRNA U79716.1 - 3 - 4 646 80

View Full Product Details