The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000108.4 | NP_000099.2 | 3 - 4 | 340 | 90 |
NM_001289752.1 | NP_001276681.1 | 3 - 4 | 340 | 90 | |
GenBank mRNA | AB209703.1 | - | 3 - 4 | 250 | 90 |
AK225289.1 | - | 3 - 4 | 264 | 90 | |
AK225301.1 | - | 3 - 4 | 276 | 90 | |
AK300077.1 | - | 3 - 4 | 273 | 90 | |
AK310220.1 | - | 3 - 4 | 261 | 90 | |
AK312346.1 | - | 3 - 4 | 287 | 90 | |
BC018648.2 | - | 3 - 4 | 249 | 90 | |
BC018696.2 | - | 3 - 4 | 249 | 90 | |
DC354209.1 | - | 3 - 4 | 340 | 90 | |
J03490.1 | - | 3 - 4 | 272 | 90 | |
J03620.1 | - | 3 - 4 | 246 | 90 |