Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NMT2
Entrez Gene ID
9397
Gene Name
N-myristoyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.10: 15104588 - 15168732 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC005232.1 - 1 - 2 128 112

View Full Product Details