Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COPZ1
Entrez Gene ID
22818
Gene Name
coatomer protein complex subunit zeta 1
Gene Aliases
CGI-120, COPZ, HSPC181, zeta-COP, zeta1-COP
Chromosome Location
Chr.12: 54325090 - 54351851 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271735.1 NP_001258664.1 2 - 3 164 74
NM_001271736.1 NP_001258665.1 2 - 3 248 74
NM_016057.2 NP_057141.1 2 - 3 164 74
NR_073424.1 2 - 3 164 74
XM_017018997.1 XP_016874486.1 2 - 3 145 74
GenBank mRNA AB047848.1 - 2 - 3 123 74
AF086911.1 - 2 - 3 111 74
AF151878.1 - 2 - 3 127 74
AF161529.1 - 2 - 3 124 74
AK025956.1 - 2 - 3 123 74
AK026190.1 - 2 - 3 123 74
AK223188.1 - 2 - 3 107 74
AK295325.1 - 2 - 3 115 74
AK311027.1 - 2 - 3 209 74
BC002849.2 - 2 - 3 106 74
CB127540.1 - 2 - 3 163 74
DA562746.1 - 2 - 3 115 74

View Full Product Details