Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COPZ1
Entrez Gene ID
22818
Gene Name
coatomer protein complex subunit zeta 1
Gene Aliases
CGI-120, COPZ, HSPC181, zeta-COP, zeta1-COP
Chromosome Location
Chr.12: 54325090 - 54351851 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271734.1 NP_001258663.1 6 - 7 453 83
NM_001271736.1 NP_001258665.1 7 - 8 606 83
NM_016057.2 NP_057141.1 7 - 8 522 83
NR_073424.1 6 - 7 430 83
GenBank mRNA AB047848.1 - 7 - 8 481 83
AF086911.1 - 7 - 8 469 83
AF151878.1 - 7 - 8 485 83
AF161529.1 - 7 - 8 482 83
AK025956.1 - 7 - 8 481 83
AK026190.1 - 7 - 8 481 83
AK223188.1 - 7 - 8 465 83
AK293377.1 - 6 - 7 404 83
AK295325.1 - 6 - 7 381 83
AK311027.1 - 7 - 8 567 83
BC002849.2 - 7 - 8 464 83

View Full Product Details