The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022104.3 | NP_071387.1 | 11 - 12 | 1479 | 94 |
XM_011528980.2 | XP_011527282.1 | 11 - 12 | 1530 | 94 | |
XM_011528981.2 | XP_011527283.1 | 11 - 12 | 1441 | 94 | |
XM_017028013.1 | XP_016883502.1 | 11 - 12 | 1640 | 94 | |
GenBank mRNA | AB050014.1 | - | 11 - 12 | 1459 | 94 |
AK056553.1 | - | - | 1457 | 94 | |
AK299435.1 | - | 11 - 12 | 1464 | 94 | |
BC010005.2 | - | 11 - 12 | 1451 | 94 | |
BC013365.2 | - | 11 - 12 | 1450 | 94 |