The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005373.3 | NP_001005373.1 | 10 - 11 | 995 | 76 |
NM_001005374.3 | NP_001005374.1 | 9 - 10 | 700 | 76 | |
NM_001190723.2 | NP_001177652.1 | 10 - 11 | 995 | 76 | |
NM_138361.5 | NP_612370.3 | 9 - 10 | 1254 | 76 | |
XM_006717316.3 | XP_006717379.1 | 10 - 11 | 995 | 76 | |
XM_017015283.1 | XP_016870772.1 | 10 - 11 | 1994 | 76 | |
XM_017015284.1 | XP_016870773.1 | 10 - 11 | 995 | 76 | |
GenBank mRNA | AK056203.1 | - | 10 - 11 | 977 | 76 |
AK091589.1 | - | 9 - 10 | 1002 | 76 | |
AK226106.1 | - | 10 - 11 | 981 | 76 | |
AY358830.1 | - | 10 - 11 | 960 | 76 | |
BC009239.2 | - | 9 - 10 | 700 | 76 |