Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
Chromosome Location
Chr.9: 127451486 - 127503501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005373.3 NP_001005373.1 20 - 21 1878 100
NM_001005374.3 NP_001005374.1 19 - 20 1583 100
NM_138361.5 NP_612370.3 19 - 20 2137 100
XM_006717316.3 XP_006717379.1 20 - 21 1878 100
XM_017015283.1 XP_016870772.1 20 - 21 2877 100
XM_017015284.1 XP_016870773.1 20 - 21 1873 100
GenBank mRNA AK056305.1 - 8 - 9 728 100
AK091589.1 - 19 - 20 1885 100
AK226106.1 - 20 - 21 1864 100
AY358830.1 - 20 - 21 1843 100
BC009239.2 - 19 - 20 1583 100

View Full Product Details