The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167865.1 | NP_001161337.1 | 5 - 6 | 814 | 89 |
NM_017615.2 | NP_060085.2 | 5 - 6 | 814 | 89 | |
XM_005269929.4 | XP_005269986.1 | 5 - 6 | 963 | 89 | |
XM_005269930.3 | XP_005269987.1 | 5 - 6 | 878 | 89 | |
XM_011539910.2 | XP_011538212.1 | 5 - 6 | 839 | 89 | |
XM_017016359.1 | XP_016871848.1 | 4 - 5 | 680 | 89 | |
GenBank mRNA | AF258584.1 | - | 4 - 5 | 1593 | 89 |
AK000010.1 | - | 5 - 6 | 781 | 89 | |
AK222487.1 | - | 5 - 6 | 781 | 89 | |
AK301658.1 | - | 5 - 6 | 912 | 89 | |
AK308687.1 | - | 6 - 7 | 857 | 89 | |
BC005212.1 | - | 5 - 6 | 780 | 89 | |
BC062427.1 | - | 5 - 6 | 807 | 89 |