The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308080.1 | NP_001295009.1 | 3 - 4 | 341 | 133 |
NM_016144.3 | NP_057228.1 | 3 - 4 | 314 | 133 | |
XM_011543458.2 | XP_011541760.1 | 3 - 4 | 698 | 133 | |
XM_011543460.2 | XP_011541762.1 | 3 - 4 | 697 | 133 | |
XM_017009553.1 | XP_016865042.1 | 3 - 4 | 436 | 133 | |
XM_017009555.1 | XP_016865044.1 | 3 - 4 | 696 | 133 | |
XM_017009556.1 | XP_016865045.1 | 3 - 4 | 696 | 133 | |
GenBank mRNA | AF078857.1 | - | 3 - 4 | 269 | 133 |
AF161423.1 | - | 3 - 4 | 265 | 133 | |
AK002147.1 | - | 3 - 4 | 290 | 133 | |
AY542165.1 | - | 3 - 4 | 237 | 133 | |
BC005179.1 | - | 3 - 4 | 260 | 133 | |
BI859871.1 | - | 3 - 4 | 330 | 133 | |
DA001572.1 | - | 3 - 4 | 341 | 133 |