Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTB
Entrez Gene ID
6710
Gene Name
spectrin beta, erythrocytic
Gene Aliases
EL3, HS2, HSPTB1, SPH2
Chromosome Location
Chr.14: 64746283 - 64879892 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000347.5 NP_000338.3 25 - 26 5603 70
NM_001024858.2 NP_001020029.1 25 - 26 5603 70
XM_005268023.4 XP_005268080.1 26 - 27 5670 70
XM_017021612.1 XP_016877101.1 27 - 28 5818 70
XM_017021613.1 XP_016877102.1 26 - 27 5670 70
XM_017021614.1 XP_016877103.1 26 - 27 5670 70
GenBank mRNA AB209415.1 - 26 - 27 5692 70
AF465439.1 - 7 - 8 1544 70
BC136284.1 - 25 - 26 5582 70
BC136285.1 - 25 - 26 5582 70
J05500.1 - 26 - 27 5644 70
M37884.1 - 11 - 12 2387 70

View Full Product Details