The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144995.1 | NP_001138467.1 | 4 - 5 | 1502 | 75 |
XM_011536705.2 | XP_011535007.1 | 5 - 6 | 1185 | 75 | |
GenBank mRNA | AK123840.1 | - | 3 - 4 | 390 | 75 |