Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDTC1
Entrez Gene ID
23038
Gene Name
WD and tetratricopeptide repeats 1
Gene Aliases
ADP, DCAF9
Chromosome Location
Chr.1: 27234516 - 27308633 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276252.1 NP_001263181.1 10 - 11 1486 87
XM_011541057.1 XP_011539359.1 10 - 11 1164 87
XM_011541058.2 XP_011539360.1 10 - 11 1431 87
GenBank mRNA AB028960.1 - 5 - 6 625 87
AK001734.1 - 9 - 10 958 87
AK315564.1 - 10 - 11 1370 87

View Full Product Details