Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDTC1
Entrez Gene ID
23038
Gene Name
WD and tetratricopeptide repeats 1
Gene Aliases
ADP, DCAF9
Chromosome Location
Chr.1: 27234516 - 27308633 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276252.1 NP_001263181.1 13 - 14 2000 122
NM_015023.4 NP_055838.2 13 - 14 1997 122
XM_011541057.1 XP_011539359.1 13 - 14 1678 122
GenBank mRNA AB028960.1 - 8 - 9 1139 122
AK001734.1 - 12 - 13 1472 122
AK023101.1 - 4 - 5 539 122
AK315564.1 - 13 - 14 1884 122
BC032523.1 - 13 - 14 1860 122

View Full Product Details