The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276252.1 | NP_001263181.1 | 3 - 4 | 662 | 57 |
NM_015023.4 | NP_055838.2 | 3 - 4 | 662 | 57 | |
XM_011541057.1 | XP_011539359.1 | 3 - 4 | 340 | 57 | |
XM_011541058.2 | XP_011539360.1 | 3 - 4 | 607 | 57 | |
XM_017000730.1 | XP_016856219.1 | 3 - 4 | 607 | 57 | |
GenBank mRNA | AK001734.1 | - | 2 - 3 | 134 | 57 |
AK315564.1 | - | 3 - 4 | 546 | 57 | |
BC032523.1 | - | 3 - 4 | 525 | 57 |