The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276252.1 | NP_001263181.1 | 5 - 6 | 823 | 106 |
NM_015023.4 | NP_055838.2 | 5 - 6 | 823 | 106 | |
XM_011541057.1 | XP_011539359.1 | 5 - 6 | 501 | 106 | |
XM_011541058.2 | XP_011539360.1 | 5 - 6 | 768 | 106 | |
XM_017000730.1 | XP_016856219.1 | 5 - 6 | 768 | 106 | |
GenBank mRNA | AK001734.1 | - | 4 - 5 | 295 | 106 |
AK315564.1 | - | 5 - 6 | 707 | 106 | |
BC032523.1 | - | 5 - 6 | 686 | 106 |