The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276252.1 | NP_001263181.1 | 6 - 7 | 1016 | 67 |
NM_015023.4 | NP_055838.2 | 6 - 7 | 1016 | 67 | |
XM_011541057.1 | XP_011539359.1 | 6 - 7 | 694 | 67 | |
XM_011541058.2 | XP_011539360.1 | 6 - 7 | 961 | 67 | |
XM_017000730.1 | XP_016856219.1 | 6 - 7 | 961 | 67 | |
GenBank mRNA | AB028960.1 | - | 1 - 2 | 155 | 67 |
AK001734.1 | - | 5 - 6 | 488 | 67 | |
AK315564.1 | - | 6 - 7 | 900 | 67 | |
BC032523.1 | - | 6 - 7 | 879 | 67 |