The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003074.3 | NP_003065.3 | 19 - 20 | 2014 | 83 |
XM_011534034.1 | XP_011532336.1 | 19 - 20 | 1745 | 83 | |
GenBank mRNA | AK302412.1 | - | 5 - 6 | 458 | 83 |
AK302541.1 | - | 19 - 20 | 1789 | 83 | |
BC021862.1 | - | 19 - 20 | 1951 | 83 | |
BC032564.1 | - | 19 - 20 | 1998 | 83 | |
BC039843.1 | - | 19 - 20 | 1918 | 83 | |
BC040242.1 | - | 4 - 5 | 446 | 83 | |
BC050564.1 | - | 19 - 20 | 1943 | 83 | |
BC065253.1 | - | 19 - 20 | 1983 | 83 | |
BC113465.1 | - | 19 - 20 | 1962 | 83 | |
BC117213.1 | - | 19 - 20 | 1962 | 83 |