The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173615.4 | NP_775886.3 | 17 - 18 | 1757 | 75 |
XM_011545742.2 | XP_011544044.1 | 20 - 21 | 1967 | 75 | |
XM_011545744.2 | XP_011544046.1 | 5 - 6 | 567 | 75 | |
XM_011545745.1 | XP_011544047.1 | 3 - 4 | 335 | 75 | |
XM_011545746.2 | XP_011544048.1 | 3 - 4 | 438 | 75 | |
XM_017022947.1 | XP_016878436.1 | 21 - 22 | 2040 | 75 | |
XM_017022948.1 | XP_016878437.1 | 4 - 5 | 1139 | 75 | |
XM_017022949.1 | XP_016878438.1 | 3 - 4 | 1105 | 75 | |
GenBank mRNA | AK128606.1 | - | 6 - 7 | 625 | 75 |
AK307544.1 | - | 18 - 19 | 1816 | 75 | |
BC109294.1 | - | 5 - 6 | 485 | 75 | |
BC109295.1 | - | 5 - 6 | 485 | 75 |