Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC19
Entrez Gene ID
131540
Gene Name
zinc finger DHHC-type containing 19
Gene Aliases
DHHC19
Chromosome Location
Chr.3: 196197443 - 196211437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039617.1 NP_001034706.1 4 - 5 689 111
NR_135617.1 4 - 5 697 111
NR_135618.1 4 - 5 697 111
XM_005269279.2 XP_005269336.1 4 - 5 697 111
XM_006713493.2 XP_006713556.1 4 - 5 667 111
XM_006713494.2 XP_006713557.1 4 - 5 595 111
XM_011512417.1 XP_011510719.1 3 - 4 454 111
GenBank mRNA AK122907.1 - 4 - 5 697 111
BC022078.2 - 4 - 5 689 111

View Full Product Details