Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PACS2
Entrez Gene ID
23241
Gene Name
phosphofurin acidic cluster sorting protein 2
Gene Aliases
PACS-2, PACS1L
Chromosome Location
Chr.14: 105300811 - 105398147 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100913.2 NP_001094383.2 14 - 15 2024 145
NM_001243127.2 NP_001230056.1 14 - 15 1534 145
NM_015197.3 NP_056012.2 14 - 15 2012 145
XM_006720090.1 XP_006720153.1 14 - 15 1692 145
XM_006720091.1 XP_006720154.1 14 - 15 1692 145
XM_006720092.2 XP_006720155.1 14 - 15 1653 145
XM_017021105.1 XP_016876594.1 14 - 15 1680 145
XM_017021106.1 XP_016876595.1 14 - 15 1677 145
XM_017021107.1 XP_016876596.1 14 - 15 1668 145
XM_017021108.1 XP_016876597.1 14 - 15 1665 145
XM_017021109.1 XP_016876598.1 14 - 15 1689 145
XM_017021110.1 XP_016876599.1 14 - 15 1680 145
XM_017021111.1 XP_016876600.1 14 - 15 1558 145
XM_017021112.1 XP_016876601.1 14 - 15 1460 145
GenBank mRNA AB011174.1 - 14 - 15 1695 145
AY320284.2 - 14 - 15 2011 145
BC010663.1 - 11 - 12 1119 145
BC050351.1 - 14 - 15 1550 145
BC065220.1 - 14 - 15 1625 145
BC131591.1 - 14 - 15 1512 145

View Full Product Details