The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100913.2 | NP_001094383.2 | 7 - 8 | 1248 | 61 |
NM_015197.3 | NP_056012.2 | 7 - 8 | 1248 | 61 | |
XM_006720090.1 | XP_006720153.1 | 7 - 8 | 913 | 61 | |
XM_006720091.1 | XP_006720154.1 | 7 - 8 | 913 | 61 | |
XM_017021105.1 | XP_016876594.1 | 7 - 8 | 913 | 61 | |
XM_017021106.1 | XP_016876595.1 | 7 - 8 | 913 | 61 | |
XM_017021109.1 | XP_016876598.1 | 7 - 8 | 913 | 61 | |
XM_017021110.1 | XP_016876599.1 | 7 - 8 | 913 | 61 | |
XM_017021111.1 | XP_016876600.1 | 7 - 8 | 794 | 61 | |
XM_017021112.1 | XP_016876601.1 | 7 - 8 | 681 | 61 | |
GenBank mRNA | AB011174.1 | - | 7 - 8 | 919 | 61 |
AY320284.2 | - | 7 - 8 | 1247 | 61 | |
BC010663.1 | - | 4 - 5 | 355 | 61 | |
BC050351.1 | - | 7 - 8 | 786 | 61 | |
BC065220.1 | - | 7 - 8 | 861 | 61 |