The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002208.4 | NP_002199.3 | 17 - 18 | 2253 | 93 |
XM_011523823.2 | XP_011522125.1 | 17 - 18 | 2691 | 93 | |
XM_011523825.2 | XP_011522127.1 | 17 - 18 | 2689 | 93 | |
XM_011523827.2 | XP_011522129.1 | 17 - 18 | 2683 | 93 | |
XM_011523828.2 | XP_011522130.1 | 17 - 18 | 2687 | 93 | |
XM_017024586.1 | XP_016880075.1 | 18 - 19 | 4076 | 93 | |
XM_017024587.1 | XP_016880076.1 | 18 - 19 | 4076 | 93 | |
GenBank mRNA | BC113436.1 | - | 17 - 18 | 93 | |
BC117207.1 | - | 17 - 18 | 93 | ||
L25851.2 | - | 17 - 18 | 93 |