The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242933.1 | NP_001229862.1 | 13 - 14 | 1574 | 70 |
NM_003099.4 | NP_003090.2 | 13 - 14 | 1574 | 70 | |
NM_148955.3 | NP_683758.1 | 11 - 12 | 1379 | 70 | |
XM_005254611.2 | XP_005254668.1 | 13 - 14 | 1550 | 70 | |
GenBank mRNA | AB209013.1 | - | 13 - 14 | 1381 | 70 |
AF065483.1 | - | 13 - 14 | 1444 | 70 | |
AF065484.1 | - | 11 - 12 | 1249 | 70 | |
AK128179.1 | - | 13 - 14 | 1466 | 70 | |
AK222561.1 | - | 13 - 14 | 1458 | 70 | |
AK222793.1 | - | 13 - 14 | 1447 | 70 | |
AK291752.1 | - | 13 - 14 | 1458 | 70 | |
BC000357.2 | - | 13 - 14 | 1454 | 70 | |
BT006983.1 | - | 13 - 14 | 1444 | 70 | |
U53225.1 | - | 13 - 14 | 1489 | 70 |