The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017615.2 | NP_060085.2 | 9 - 10 | 1147 | 144 |
XM_005269929.4 | XP_005269986.1 | 9 - 10 | 1296 | 144 | |
XM_005269930.3 | XP_005269987.1 | 9 - 10 | 1211 | 144 | |
XM_011539910.2 | XP_011538212.1 | 9 - 10 | 1172 | 144 | |
XM_017016359.1 | XP_016871848.1 | 8 - 9 | 1013 | 144 | |
GenBank mRNA | AF258584.1 | - | 9 - 10 | 2193 | 144 |
AK000010.1 | - | 9 - 10 | 1114 | 144 | |
AK222487.1 | - | 9 - 10 | 1114 | 144 |